COMH4211B06E Combi-Blocks, Inc.

Gedunin

  • MDL number MFCD01750174
  • Cas Number : 2753-30-2
  • Molecular Weight : 482.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3527
Availability details for
Quantity
  • Availability
  • Add to Cart