COMH4211B06F Combi-Blocks, Inc.

Pentanoic acid, 5-[(3-iodobenzoyl)amino]-

  • MDL number MFCD19450385
  • Cas Number : 1458544-53-0
  • Molecular Weight : 347.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3528
Availability details for
Quantity
  • Availability
  • Add to Cart