COMH4211B072 Combi-Blocks, Inc.

3-(Propane-1-sulfonyl)-propionic acid

  • MDL number MFCD09951319
  • Cas Number : 99116-16-2
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3535
Availability details for
Quantity
  • Availability
  • Add to Cart