COMH4211B075 Combi-Blocks, Inc.

1-(4-Ethoxy-2-fluoro-5-nitro-phenyl)-ethanone

  • MDL number MFCD31613710
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3539
Availability details for
Quantity
  • Availability
  • Add to Cart