COMH4211B079 Combi-Blocks, Inc.

2,2-Dimethyl-3-(pyrrolidin-1-yl)propanoic acid hydrochloride

  • MDL number MFCD31381681
  • Cas Number : 2206610-48-0
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3553
Availability details for
Quantity
  • Availability
  • Add to Cart