COMH4211B07C Combi-Blocks, Inc.

(3-Chloro-2-cyclopropylmethoxy-benzyl)-methyl-amine

  • MDL number MFCD25339379
  • Cas Number : 1565444-22-5
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3556

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3556
Availability details for
Quantity
  • Availability
  • Add to Cart