COMH4211B07D Combi-Blocks, Inc.

4-(2,2,2-Trifluoroethoxy)-2-trifluoromethylphenylamine

  • MDL number MFCD11132422
  • Cas Number : 1019441-72-5
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3557
Availability details for
Quantity
  • Availability
  • Add to Cart