COMH4211B07F Combi-Blocks, Inc.

4-[(1,3-Dioxo-1,3-dihydro-2h-isoindol-2-yl)oxy]methylbenzonitrile

  • MDL number MFCD31614048
  • Cas Number : 2086-27-3
  • Molecular Weight : 278.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3562

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3562
Availability details for
Quantity
  • Availability
  • Add to Cart