COMH4211B081 Combi-Blocks, Inc.

3-(4-(4-Chlorophenyl)piperazin-1-yl)propanoic acid

  • MDL number MFCD11537367
  • Cas Number : 732298-42-9
  • Molecular Weight : 268.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3565

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3565
Availability details for
Quantity
  • Availability
  • Add to Cart