COMH4211B083 Combi-Blocks, Inc.

3-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenoxymethyl]-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583289
  • Molecular Weight : 417.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3569
Availability details for
Quantity
  • Availability
  • Add to Cart