COMH4211B085 Combi-Blocks, Inc.

2-Cyclopentylsulfanyl-3-iodo-pyridine

  • MDL number MFCD31539533
  • Cas Number : 1643457-06-0
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3574
Availability details for
Quantity
  • Availability
  • Add to Cart