COMH4211B087 Combi-Blocks, Inc.

2-Amino-3-(4-fluoro-2-methoxy-phenyl)-propionic acid hydrochloride

  • MDL number MFCD31561582
  • Cas Number : 2270904-96-4
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3576
Availability details for
Quantity
  • Availability
  • Add to Cart