COMH4211B088 Combi-Blocks, Inc.

[2-(4-Hydroxy-pyrazol-1-yl)-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD31579325
  • Cas Number : 2270908-91-1
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3578
Availability details for
Quantity
  • Availability
  • Add to Cart