COMH4211B08A Combi-Blocks, Inc.

3-(6-Methoxynaphthalen-2-yl)propanoic acid

  • MDL number MFCD09940475
  • Cas Number : 3453-40-5
  • Molecular Weight : 230.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3591

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3591
Availability details for
Quantity
  • Availability
  • Add to Cart