COMH4211B08B Combi-Blocks, Inc.

2-(5-Fluoropyrimidin-2-yl)benzonitrile

  • MDL number MFCD30470792
  • Cas Number : 1637687-38-7
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3593
Availability details for
Quantity
  • Availability
  • Add to Cart