COMH4211B08C Combi-Blocks, Inc.

2-Bromo-6-(cyclohexyloxy)benzonitrile

  • MDL number MFCD28129645
  • Cas Number : 1369954-21-1
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3594

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3594
Availability details for
Quantity
  • Availability
  • Add to Cart