COMH4211B08E Combi-Blocks, Inc.

Methyl 4-((4-bromobenzyl)carbamoyl)benzoate

  • MDL number MFCD31614043
  • Cas Number : 2052958-39-9
  • Molecular Weight : 348.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3601
Availability details for
Quantity
  • Availability
  • Add to Cart