COMH4211B08F Combi-Blocks, Inc.

4-(4-Formylphenoxy)butanoic acid

  • MDL number MFCD02258439
  • Cas Number : 99865-70-0
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-formylphenoxy)butanoic acid
Inchi Code 1S/C11H12O4/c12-8-9-3-5-10(6-4-9)15-7-1-2-11(13)14/h3-6,8H,1-2,7H2,(H,13,14)
Inchi Key GKECISRCTNHSNA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3604
Availability details for
Quantity
  • Availability
  • Add to Cart