COMH4211B090 Combi-Blocks, Inc.

3-Fluoro-4-isobutoxybenzoic acid

  • MDL number MFCD11156597
  • Cas Number : 1021144-45-5
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3606

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3606
Availability details for
Quantity
  • Availability
  • Add to Cart