COMH4211B093 Combi-Blocks, Inc.

4-(2-Fluoro-4-isopropoxy-phenyl)-1,2,3,6-tetrahydro-pyridine hydrochloride

  • MDL number MFCD31583385
  • Cas Number : 2270905-41-2
  • Molecular Weight : 271.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-fluoro-4-isopropoxyphenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Inchi Code 1S/C14H18FNO.ClH/c1-10(2)17-12-3-4-13(14(15)9-12)11-5-7-16-8-6-11;/h3-5,9-10,16H,6-8H2,1-2H3;1H
Inchi Key KXJOTPYOHUYLLK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3612
Availability details for
Quantity
  • Availability
  • Add to Cart