COMH4211B094 Combi-Blocks, Inc.

5-Bromo-2-(2-methoxy-ethoxy)-benzamide

  • MDL number MFCD07628286
  • Cas Number : 130840-20-9
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3618
Availability details for
Quantity
  • Availability
  • Add to Cart