COMH4211B097 Combi-Blocks, Inc.

1-Difluoromethoxy-4-ethynyl-2-methoxy-benzene

  • MDL number MFCD31557817
  • Cas Number : 1062617-51-9
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3622

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3622
Availability details for
Quantity
  • Availability
  • Add to Cart