COMH4211B098 Combi-Blocks, Inc.

(4-Nitro-cyclohex-3-enyl)-carbamic acid tert-butyl ester

  • MDL number MFCD31614116
  • Cas Number : 2270906-00-6
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3623
Availability details for
Quantity
  • Availability
  • Add to Cart