COMH4211B09E Combi-Blocks, Inc.

3-Chloro-4-vinylpyridine

  • MDL number MFCD22190538
  • Cas Number : 1168722-58-4
  • Molecular Weight : 139.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3633
Availability details for
Quantity
  • Availability
  • Add to Cart