COMH4211B0A1 Combi-Blocks, Inc.

3-Bromo-n,n-dimethyl-5-(propane-2-sulfonylamino)-benzamide

  • MDL number MFCD31613982
  • Cas Number : 2270913-24-9
  • Molecular Weight : 349.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3642

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3642
Availability details for
Quantity
  • Availability
  • Add to Cart