COMH4211B0A5 Combi-Blocks, Inc.

(1,3-Dihydro-isoindol-2-yl)-(4-iodo-phenyl)-methanone

  • MDL number MFCD17946175
  • Cas Number : 1283764-23-7
  • Molecular Weight : 349.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3647
Availability details for
Quantity
  • Availability
  • Add to Cart