COMH4211B0A6 Combi-Blocks, Inc.

3-(4-Amino-phenylsulfanylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD25973702
  • Cas Number : 1432059-89-6
  • Molecular Weight : 322.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3650

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3650
Availability details for
Quantity
  • Availability
  • Add to Cart