COMH4211B0A7 Combi-Blocks, Inc.

2,2'-Dichlorobiphenyl-4-amine

  • MDL number MFCD27940997
  • Cas Number : 102871-33-0
  • Molecular Weight : 238.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3656
Availability details for
Quantity
  • Availability
  • Add to Cart