COMH4211B0A8 Combi-Blocks, Inc.

2-[3,5-Dimethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-ylmethyl]-benzonitrile

  • MDL number MFCD31583209
  • Cas Number : 2246901-96-0
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3657
Availability details for
Quantity
  • Availability
  • Add to Cart