COMH4211B0A9 Combi-Blocks, Inc.

Benzene, 1-bromo-3-chloro-2-[[(4-methoxyphenyl)methoxy]methyl]-

  • MDL number MFCD31613867
  • Cas Number : 1980784-00-6
  • Molecular Weight : 341.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3658
Availability details for
Quantity
  • Availability
  • Add to Cart