COMH4211B0AA Combi-Blocks, Inc.

4-(4-Vinyl-phenoxy)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD11215289
  • Cas Number : 1146080-20-7
  • Molecular Weight : 303.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3661

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3661
Availability details for
Quantity
  • Availability
  • Add to Cart