COMH4211B0AC Combi-Blocks, Inc.

6-(4-Isopropylpiperazin-1-yl)pyridin-3-amine

  • MDL number MFCD11044160
  • Cas Number : 837421-41-7
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3664
Availability details for
Quantity
  • Availability
  • Add to Cart