COMH4211B0AE Combi-Blocks, Inc.

2-[(3-Methylpiperidin-1-yl)methyl]aniline

  • MDL number MFCD09809204
  • Cas Number : 1016679-71-2
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3667
Availability details for
Quantity
  • Availability
  • Add to Cart