COMH4211B0AF Combi-Blocks, Inc.

4-(tert-Butyldimethylsilyloxy)-3-methylaniline

  • MDL number MFCD26397178
  • Cas Number : 309712-95-6
  • Molecular Weight : 237.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3668
Availability details for
Quantity
  • Availability
  • Add to Cart