COMH4211B0B0 Combi-Blocks, Inc.

2-(4,4-Dimethylcyclohexyl)ethan-1-ol

  • MDL number MFCD31390972
  • Cas Number : 149394-69-4
  • Molecular Weight : 156.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3669

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3669
Availability details for
Quantity
  • Availability
  • Add to Cart