COMH4211B0C7 Combi-Blocks, Inc.

1H-Indene, 5-(3-bromopropoxy)-2,3-dihydro-

  • MDL number MFCD11164583
  • Cas Number : 78114-95-1
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3674
Availability details for
Quantity
  • Availability
  • Add to Cart