COMH4211B0C9 Combi-Blocks, Inc.

N-(2-[2-(2-Amino-ethoxy)-ethoxy]-ethyl)-4-sulfamoyl-benzamide

  • MDL number MFCD30578229
  • Cas Number : 165618-72-4
  • Molecular Weight : 331.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3677
Availability details for
Quantity
  • Availability
  • Add to Cart