COMH4211B0CB Combi-Blocks, Inc.

1-(3-Chloro-4-propoxyphenyl)ethan-1-amine

  • MDL number MFCD15175427
  • Cas Number : 752924-30-4
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3679
Availability details for
Quantity
  • Availability
  • Add to Cart