COMH4211B0CD Combi-Blocks, Inc.

C-(5-Chloro-2-cyclopropylmethoxy-phenyl)-c-phenyl-methylamine hydrochloride

  • MDL number MFCD31557840
  • Cas Number : 2208788-16-1
  • Molecular Weight : 324.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3690

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3690
Availability details for
Quantity
  • Availability
  • Add to Cart