COMH4211B0D1 Combi-Blocks, Inc.

N-Depropyl propafenone oxalate salt

  • MDL number MFCD09840339
  • Cas Number : 86383-21-3
  • Molecular Weight : 389.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[2-(3-amino-2-hydroxypropoxy)phenyl]-3-phenyl-1-propanone oxalate
Inchi Code 1S/C18H21NO3.C2H2O4/c19-12-15(20)13-22-18-9-5-4-8-16(18)17(21)11-10-14-6-2-1-3-7-14;3-1(4)2(5)6/h1-9,15,20H,10-13,19H2;(H,3,4)(H,5,6)
Inchi Key UMGRCTLZIBOZHC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3698
Availability details for
Quantity
  • Availability
  • Add to Cart