COMH4211B0D3 Combi-Blocks, Inc.

2-Chloro-n-cyclobutylpyridin-4-amine

  • MDL number MFCD23924800
  • Cas Number : 1289208-41-8
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-N-cyclobutylpyridin-4-amine
Inchi Code 1S/C9H11ClN2/c10-9-6-8(4-5-11-9)12-7-2-1-3-7/h4-7H,1-3H2,(H,11,12)
Inchi Key JRDRXCLFCGYEOG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3700
Availability details for
Quantity
  • Availability
  • Add to Cart