COMH4211B0D5 Combi-Blocks, Inc.

3-[2-(2-(2-[2-(Toluene-4-sulfonyloxy)-ethoxy]-ethoxy)-ethoxy)-ethoxy]-propionic acid

  • MDL number MFCD31381633
  • Cas Number : 2205504-91-0
  • Molecular Weight : 420.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3702

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3702
Availability details for
Quantity
  • Availability
  • Add to Cart