COMH4211B0D6 Combi-Blocks, Inc.

5-Amino-4-fluoro-n-isopropyl-2-methyl-benzamide

  • MDL number MFCD31579290
  • Cas Number : 1871156-43-2
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3713
Availability details for
Quantity
  • Availability
  • Add to Cart