COMH4211B0D7 Combi-Blocks, Inc.

N-Cyclopropyl-1-(3-bromophenyl)-1,4-dihydro[1,8]naphthyridin-4-one-3-carboxamide

  • MDL number MFCD31579566
  • Cas Number : 477251-96-0
  • Molecular Weight : 384.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3715
Availability details for
Quantity
  • Availability
  • Add to Cart