COMH4211B0D9 Combi-Blocks, Inc.

1-(4-Bromo-2-fluorophenyl)cyclobutanecarbonitrile

  • MDL number MFCD19697275
  • Cas Number : 749929-04-2
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3718

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3718
Availability details for
Quantity
  • Availability
  • Add to Cart