COMH4211B0DA Combi-Blocks, Inc.

4'-Bromo-3'-methyl-biphenyl-3-carboxylic acid

  • MDL number MFCD31579462
  • Cas Number : 2270905-54-7
  • Molecular Weight : 291.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3720
Availability details for
Quantity
  • Availability
  • Add to Cart