COMH4211B0DD Combi-Blocks, Inc.

1-(3-Chloro-2-cyclopropylmethoxy-phenyl)-ethylamine

  • MDL number MFCD31557800
  • Cas Number : 1784125-69-4
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3728
Availability details for
Quantity
  • Availability
  • Add to Cart