COMH4211B0DF Combi-Blocks, Inc.

C-Phenyl-c-(4-trifluoromethoxy-phenyl)-methylamine hydrochloride

  • MDL number MFCD31561548
  • Cas Number : 49703-68-6
  • Molecular Weight : 303.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3731
Availability details for
Quantity
  • Availability
  • Add to Cart