COMH4211B0E1 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-[2-(4-nitro-benzyloxy)-phenyl]-[1,3,2]dioxaborolane

  • MDL number MFCD31614094
  • Molecular Weight : 355.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3735
Availability details for
Quantity
  • Availability
  • Add to Cart