COMH4211B0E3 Combi-Blocks, Inc.

4-Bromo-1-(cyclopropylmethoxy)-2-methoxybenzene

  • MDL number MFCD29095068
  • Cas Number : 1369875-57-9
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3741
Availability details for
Quantity
  • Availability
  • Add to Cart